Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Loxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013586.4 | NP_038614.2 | 2, 3 | 417 | ||
GenBank mRNA | AF053368.1 | - | 2, 3 | 448 | ||
AK030548.1 | - | 2, 3 | 418 | |||
AK145189.1 | - | 2, 3 | 437 | |||
BC011298.1 | - | 2, 3 | 411 | 3488978 |