Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lum | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008524.2 | NP_032550.2 | 3 | 1309 | ||
GenBank mRNA | AK014312.1 | - | 3 | 1005 | ||
AK075737.1 | - | 3 | 1008 | |||
AK085240.2 | - | 3 | 1001 | |||
AK164774.1 | - | 3 | 1001 | |||
BC005550.1 | - | 3 | 987 | 3585672 | ||
S79461.1 | - | 3 | 990 |