Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marcks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008538.2 | NP_032564.1 | 2 | 1943 | ||
GenBank mRNA | AK082623.1 | - | 1 | 94 | ||
AK136452.1 | - | 1 | 112 | |||
AK151646.1 | - | 1 | 379 | |||
BC046601.1 | - | 2 | 1904 | 6311439 | ||
M60474.1 | - | 2 | 1933 | |||
X59374.1 | - | 2 | 1834 |