Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Madcam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013591.2 | NP_038619.2 | 4 | 741 | ||
GenBank mRNA | AK137028.1 | - | 4 | 742 | ||
BE632565.1 | - | 2 | 84 | |||
L21203.1 | - | 4 | 738 |