Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164041.1 | NP_001157513.1 | 5 | 1632 | ||
NM_001164042.1 | NP_001157514.1 | 5 | 1252 | |||
NM_008541.3 | NP_032567.1 | 5 | 1229 | |||
GenBank mRNA | AF063006.1 | - | 5 | 1633 | ||
AK018077.1 | - | 5 | 1201 | |||
AK082997.1 | - | 5 | 1253 | |||
AK139765.1 | - | 6 | 1277 | |||
BC050001.1 | - | 5 | 1132 | 6490853 | ||
U58993.1 | - | 5 | 1208 | |||
U77638.1 | - | 5 | 1378 |