Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025577.2 | NP_001020748.2 | 1 | 103 | ||
GenBank mRNA | AK132165.1 | - | 82 | |||
AK151738.1 | - | 244 | ||||
AK152074.1 | - | 244 | ||||
S74567.1 | - | 1003 |