Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010764.2 | NP_034894.2 | 16 | 2064 | ||
GenBank mRNA | AK004817.2 | - | 16 | 2021 | ||
AK035551.1 | - | 16 | 2018 | |||
AK147928.1 | - | 16 | 2022 | |||
AK150154.1 | - | 13 | 1396 | |||
AK170848.1 | - | 13 | 1395 | |||
AK171166.1 | - | 16 | 2022 | |||
AK171377.1 | - | 13 | 1395 | |||
BC005430.1 | - | 16 | 2009 | 3488681 | ||
BC023332.1 | - | 7 | 766 | |||
BC035318.1 | - | 3 | 279 | |||
U29947.1 | - | 16 | 2007 | |||
U87240.1 | - | 16 | 1998 |