Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314039.1 | NP_001300968.1 | 2 | 298 | ||
NM_001539.3 | NP_001530.1 | 2 | 298 | |||
GenBank mRNA | AK289623.1 | - | 2 | 226 | ||
AK298725.1 | - | 2 | 226 | |||
AY186741.1 | - | 2 | 224 | |||
BC008182.1 | - | 2 | 211 | 2900892 | ||
BI462555.1 | - | 2 | 293 | |||
BM469197.1 | - | 2 | 199 | |||
BT007292.1 | - | 1 | 107 | |||
D13388.1 | - | 2 | 224 | |||
DA003255.1 | - | 2 | 300 | |||
L08069.1 | - | 2 | 189 |