Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008576.3 | NP_032602.1 | 23 | 3246 | ||
GenBank mRNA | AF022908.1 | - | 23 | 3208 | ||
AK029876.1 | - | 23 | 3247 | |||
AK033750.1 | - | 1 | 29 | |||
AK049174.1 | - | 7 | 1013 | |||
AK140421.1 | - | 1 | 23 | |||
BC090617.1 | - | 23 | 3217 | 30943198 |