Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021557.5 | NP_067532.2 | 5, 6 | 693 | ||
GenBank mRNA | AB030503.1 | - | 5, 6 | 1025 | ||
AB030504.1 | - | 5, 6 | 773 | |||
AB035959.1 | - | 5, 6 | 701 | |||
AF474027.1 | - | 5, 6 | 739 | |||
AK004413.1 | - | 5, 6 | 694 | |||
AK012056.1 | - | 5, 6 | 721 | |||
AK135443.1 | - | 5, 6 | 723 | |||
AY039032.1 | - | 5, 6 | 1167 | |||
BC018261.1 | - | 5, 6 | 718 | 4460226 | ||
BC036273.1 | - | 5, 6 | 718 |