Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023126.2 | NP_075615.2 | 4 | 467 | ||
GenBank mRNA | AB232600.1 | - | 4 | 308 | ||
AK009259.1 | - | 4 | 318 | |||
AK010862.1 | - | 4 | 318 | |||
AK076048.1 | - | 4 | 346 | |||
AK079306.1 | - | 334 | ||||
AK080740.1 | - | 4 | 319 | |||
AK147178.1 | - | 4 | 335 | |||
AK165690.1 | - | 4 | 330 | |||
AY974570.1 | - | 4 | 286 | |||
BC019990.1 | - | 4 | 308 | 4161110 | ||
S53270.1 | - | 4 | 325 |