Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008597.3 | NP_032623.1 | 2 | 152 | ||
GenBank mRNA | AK018719.1 | - | 2 | 153 | ||
AK159591.1 | - | 2 | 156 | |||
BC018338.1 | - | 2 | 44 | |||
BC036991.1 | - | 2 | 148 | 4990627 | ||
BC069922.1 | - | 2 | 142 | 4984262 | ||
BC079478.1 | - | 2 | 143 | 4950210 | ||
BY702829.1 | - | 2 | 153 | |||
D00613.1 | - | 2 | 95 |