Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252560.1 | NP_001239489.1 | 7 | 839 | ||
NM_001252561.1 | NP_001239490.1 | 8 | 890 | |||
NM_010804.4 | NP_034934.2 | 7 | 694 | |||
GenBank mRNA | AF010135.1 | - | 7 | 737 | ||
AK045042.1 | - | 7 | 594 | |||
AK147388.1 | - | 10 | 1184 | |||
AK154320.1 | - | 7 | 839 | |||
AK158426.1 | - | 6 | 432 | |||
AK220504.1 | - | 9 | 946 | |||
BC055298.1 | - | 8 | 839 | |||
BC069988.1 | - | 6 | 688 | |||
BC070475.1 | - | 7 | 660 | 30638433 | ||
BY608946.1 | - | 2 | 138 | |||
CN534499.1 | - | 8 | 727 | |||
CN537566.1 | - | 4 | 367 |