Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252560.1 | NP_001239489.1 | 9 | 1061 | ||
NM_001252561.1 | NP_001239490.1 | 10 | 1112 | |||
NM_010804.4 | NP_034934.2 | 9 | 916 | |||
GenBank mRNA | AK045042.1 | - | 9 | 817 | ||
AK147388.1 | - | 12 | 1406 | |||
AK154320.1 | - | 9 | 1061 | |||
AK158426.1 | - | 8 | 654 | |||
AK220504.1 | - | 11 | 1168 | |||
BC055298.1 | - | 10 | 1061 | |||
BC070475.1 | - | 9 | 882 | 30638433 | ||
CN537566.1 | - | 6 | 589 |