Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010808.3 | NP_034938.3 | 8 | 1397 | ||
GenBank mRNA | AB021226.1 | - | 8 | 1342 | ||
AJ010262.1 | - | 8 | 1340 | |||
BM950641.1 | - | 2 | 140 | |||
CD352405.1 | - | 5 | 594 |