Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008612.2 | NP_032638.2 | 7 | 823 | ||
GenBank mRNA | AK008134.1 | - | 7 | 788 | ||
AK008249.1 | - | 7 | 826 | |||
AK011833.1 | - | 6 | 710 | |||
AK029174.1 | - | 7 | 735 | |||
AK155859.1 | - | 7 | 824 | |||
AK157721.1 | - | 7 | 810 | |||
BC089023.1 | - | 7 | 812 | 30702121 | ||
BC115630.2 | - | 7 | 815 | 40102880 | ||
U35249.1 | - | 7 | 854 |