Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039368.2 | NP_001034457.1 | 3 | 330 | ||
NM_023127.3 | NP_075616.2 | 3 | 198 | |||
GenBank mRNA | AK003023.1 | - | 3 | 180 | ||
AK008136.1 | - | 3 | 287 | |||
AK008287.1 | - | 3 | 288 | |||
AK018914.1 | - | 3 | 157 | |||
BC028543.1 | - | 146 | 3468916 | |||
BY100585.1 | - | 3 | 193 | |||
BY646564.1 | - | 2 | 122 | |||
S63758.1 | - | 3 | 259 |