Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173506.1 | NP_001166977.1 | 5 | 8469 | ||
NM_032393.2 | NP_115769.1 | 6 | 8753 | |||
GenBank mRNA | AK147474.1 | - | 2 | 6965 | ||
AK163468.1 | - | 2 | 354 |