Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198914.1 | NP_001185843.1 | 5 | 679 | ||
NM_010848.3 | NP_034978.3 | 5 | 679 | |||
GenBank mRNA | AK088079.1 | - | 5 | 600 | ||
AK088598.1 | - | 5 | 601 | |||
AK132250.1 | - | 5 | 544 | |||
AK137965.1 | - | 5 | 545 | |||
BC011513.1 | - | 5 | 587 | 3672769 | ||
M12848.1 | - | 5 | 677 | |||
X02774.1 | - | 5 | 449 |