Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201600.2 | NP_963894.1 | 36, 37 | 5210 | ||
GenBank mRNA | AK033484.1 | - | 6, 7 | 1063 | ||
AK173102.1 | - | 13, 14 | 4664 | |||
BC011494.1 | - | 15, 16 | 2169 | |||
BC046444.1 | - | 27, 28 | 3919 | |||
BC057910.1 | - | 7, 8 | 1250 | |||
BC118525.1 | - | 36, 37 | 5203 | 40099275 | ||
M55253.1 | - | 13, 14 | 1809 |