Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008677.2 | NP_032703.2 | 4 | 528 | ||
GenBank mRNA | AB002665.1 | - | 4 | 505 | ||
AK150510.1 | - | 4 | 510 | |||
AK170854.1 | - | 1 | 36 | |||
AK171315.1 | - | 4 | 306 | |||
BC025517.1 | - | 4 | 357 | 5321182 | ||
FJ168470.1 | - | 4 | 319 | |||
U59488.1 | - | 4 | 475 |