Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008682.2 | NP_032708.2 | 4 | 424 | ||
GenBank mRNA | AK010591.1 | - | 4 | 412 | ||
AK048393.1 | - | 4 | 376 | |||
AK085355.1 | - | 4 | 387 | |||
AK090040.1 | - | 4 | 369 | |||
AK153746.1 | - | 4 | 426 | |||
BC052430.1 | - | 4 | 393 | 6836583 | ||
BC066870.1 | - | 4 | 361 | 30476383 | ||
D10712.1 | - | 4 | 366 |