Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neurod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010894.2 | NP_035024.1 | 2 | 507 | ||
GenBank mRNA | AK005073.1 | - | 2 | 508 | ||
AK018781.1 | - | 2 | 506 | |||
AK028055.1 | - | 2 | 511 | |||
BC018241.1 | - | 2 | 483 | 4511370 | ||
BC094611.1 | - | 2 | 447 | 30361598 | ||
U28068.1 | - | 1 | 194 |