Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010897.2 | NP_035027.1 | 38 | 5471 | ||
GenBank mRNA | L10370.1 | - | 37 | 5316 |