Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008694.2 | NP_032720.2 | 3, 4 | 413 | ||
GenBank mRNA | AK009649.1 | - | 3, 4 | 367 | ||
AK165210.1 | - | 3, 4 | 366 | |||
BC119403.1 | - | 3, 4 | 384 | 8734152 | ||
BC119405.1 | - | 3, 4 | 384 | 8734154 | ||
L37297.1 | - | 3, 4 | 418 | |||
U95002.1 | - | 3, 4 | 372 |