Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008695.2 | NP_032721.2 | 4 | 1540 | ||
GenBank mRNA | AB017202.1 | - | 4 | 1502 | ||
AK045306.1 | - | 4 | 1515 | |||
AK051664.1 | - | 4 | 1516 | |||
AK052839.1 | - | 4 | 1514 | |||
AK083629.1 | - | 4 | 1516 | |||
AK128978.1 | - | 1 | 662 | |||
AK140832.1 | - | 4 | 1514 | |||
AK164258.1 | - | 4 | 1511 | |||
AY074820.1 | - | 4 | 1501 | |||
BC054746.1 | - | 4 | 1522 | 6405073 | ||
BC057016.1 | - | 4 | 1468 | 5719452 |