Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M24630.1 | - | 15 | 3732 | ||
M55618.1 | - | 16 | 5157 | |||
X78565.1 | - | 17 | 5335 |