Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc11a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013612.2 | NP_038640.2 | 8, 9 | 900 | ||
GenBank mRNA | AK090303.1 | - | 8, 9 | 901 | ||
AK144237.1 | - | 8, 9 | 884 | |||
AK171343.1 | - | 7, 8 | 1136 | |||
AK171393.1 | - | 8, 9 | 881 | |||
BC109137.1 | - | 8, 9 | 814 | 40039390 | ||
BC109138.1 | - | 8, 9 | 814 | 40039397 | ||
L13732.1 | - | 8, 9 | 878 | |||
X75355.1 | - | 8, 9 | 859 |