Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsdhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010941.3 | NP_035071.3 | 3 | 370 | ||
GenBank mRNA | AF100198.1 | - | 3 | 370 | ||
AK013945.1 | - | 3 | 253 | |||
AK088026.1 | - | 3 | 253 | |||
AK140930.1 | - | 3 | 251 | |||
BC019945.1 | - | 3 | 239 | 3989811 | ||
BC052834.1 | - | 3 | 331 | 30048305 | ||
KF356400.1 | - | 2 | 155 | |||
KF356401.1 | - | 2 | 155 |