Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008750.5 | NP_032776.1 | 1, 2 | 435 | ||
GenBank mRNA | AB095441.1 | - | 1, 2 | 280 | ||
AB095442.1 | - | 1, 2 | 280 | |||
AB095443.1 | - | 1, 2 | 280 | |||
AB095444.1 | - | 1, 2 | 280 | |||
AK012352.1 | - | 1, 2 | 420 | |||
AK048109.1 | - | 1, 2 | 434 | |||
AK149075.1 | - | 1, 2 | 419 | |||
BC004688.1 | - | 1, 2 | 395 | 3501314 | ||
BC058244.1 | - | 1, 2 | 435 | 6312910 |