Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113213.1 | NP_001106684.1 | 8 | 1103 | ||
NM_001113214.1 | NP_001106685.1 | 8 | 1103 | |||
NM_001177659.1 | NP_001171130.1 | 8 | 1103 | |||
NM_001177661.1 | NP_001171132.1 | 7 | 949 | |||
NM_013615.3 | NP_038643.1 | 8 | 1076 | |||
GenBank mRNA | AF000968.1 | - | 6 | 923 | ||
AF034105.1 | - | 8 | 1039 | |||
AK143726.1 | - | 8 | 1007 | |||
AK148523.1 | - | 8 | 1016 | |||
AK165588.1 | - | 8 | 1007 | |||
AK168415.1 | - | 8 | 1005 | |||
BC057001.1 | - | 9 | 1160 | 6827260 |