Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146563.1 | NP_666774.1 | 2 | 696 | ||
GenBank mRNA | BC120866.1 | - | 2 | 595 | 40129789 | |
BC120868.1 | - | 2 | 595 | 40129791 | ||
BC144957.1 | - | 3 | 606 | |||
X89686.1 | - | 1 | 127 |