Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146946.1 | NP_667157.1 | 1 | 841 | ||
GenBank mRNA | AF102524.1 | - | 1 | 652 | ||
BC127978.1 | - | 1 | 845 | 40135208 |