Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Omt2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111286.2 | NP_001104756.1 | 2 | 765 | ||
NM_001164523.1 | NP_001157995.1 | 2 | 144 | |||
GenBank mRNA | AA619817.1 | - | 2 | 133 | ||
AA693017.1 | - | 2 | 144 | |||
AK135846.1 | - | 2 | 191 | |||
AK139546.1 | - | 2 | 733 | |||
BB698495.1 | - | 2 | 75 | |||
DV647767.1 | - | 2 | 197 | |||
DV648064.1 | - | 2 | 209 | |||
S81934.1 | - | 3 | 301 |