Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Omt2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111286.2 | NP_001104756.1 | 2 | 813 | ||
NM_001164523.1 | NP_001157995.1 | 2 | 192 | |||
GenBank mRNA | AA619817.1 | - | 2 | 181 | ||
AA693017.1 | - | 2 | 192 | |||
AK135846.1 | - | 2 | 239 | |||
AK139546.1 | - | 2 | 781 | |||
BB698495.1 | - | 2 | 123 | |||
DV647767.1 | - | 2 | 245 | |||
DV648064.1 | - | 2 | 257 | |||
S81934.1 | - | 3 | 349 |