Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ovol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019935.3 | NP_064319.1 | 3 | 770 | ||
GenBank mRNA | AF134804.1 | - | 3 | 786 | ||
AF134805.1 | - | 3 | 752 | |||
AK003801.1 | - | 3 | 805 | |||
AK028577.1 | - | 3 | 771 | |||
AK132478.1 | - | 3 | 771 | |||
AK132505.1 | - | 3 | 809 | |||
AK132506.1 | - | 3 | 805 | |||
AK144952.1 | - | 3 | 805 | |||
BC021411.1 | - | 3 | 850 | 5006712 |