Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021879.2 | NP_068679.1 | 21, 22 | 2363 | ||
GenBank mRNA | AK051897.1 | - | 20, 21 | 2204 | ||
AK087494.1 | - | 22, 23 | 2623 | |||
BC119220.1 | - | 21, 22 | 2333 | 8733969 | ||
BC120549.1 | - | 21, 22 | 2333 | 40129472 | ||
M97900.1 | - | 21, 22 | 2363 |