Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190846.1 | NP_001177775.1 | 14 | 1792 | ||
NM_020005.4 | NP_064389.2 | 14 | 2346 | |||
GenBank mRNA | AF254442.1 | - | 14 | 1987 | ||
AK030070.1 | - | 14 | 1793 | |||
AK156290.1 | - | 14 | 2346 | |||
BC019722.1 | - | 2 | 152 | |||
BC082581.1 | - | 14 | 2002 | 30653109 | ||
BC138195.1 | - | 14 | 2014 | 8861214 | ||
BC145896.1 | - | 14 | 2014 | 40131199 |