Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282992.1 | NP_001269921.1 | 22 | 4290 | ||
NM_008787.3 | NP_032813.3 | 22 | 4290 | |||
GenBank mRNA | AB207233.1 | - | 22 | 4164 | ||
AB207234.1 | - | 10 | 1585 | |||
AK122275.2 | - | 7 | 1037 | |||
BC100380.1 | - | 22 | 4085 |