Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF339910.1 | - | 15 | 1457 | ||
AK029600.1 | - | 15 | 1369 | |||
AK036952.1 | - | 15 | 1466 | |||
AK041894.1 | - | 10 | 830 | |||
AK088857.1 | - | 15 | 1464 | |||
AK145233.1 | - | 15 | 1460 | |||
AK164619.1 | - | 15 | 1439 | |||
BC050011.1 | - | 15 | 1611 | |||
J02700.2 | - | 15 | 1568 |