Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pepd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008820.2 | NP_032846.2 | 7 | 546 | ||
GenBank mRNA | AK010581.1 | - | 7 | 527 | ||
AK145843.1 | - | 7 | 577 | |||
AK168384.1 | - | 7 | 536 | |||
AK206317.1 | - | 2 | 72 | |||
BC086644.1 | - | 7 | 520 | 6807359 | ||
BC131945.1 | - | 7 | 548 | 40130222 | ||
BC131973.1 | - | 7 | 548 | 40130250 | ||
D82983.1 | - | 7 | 533 | |||
U51014.1 | - | 7 | 569 |