Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162387.1 | NP_001155859.1 | 4 | 661 | ||
NM_001162388.1 | NP_001155860.1 | 4 | 425 | |||
NM_011069.3 | NP_035199.3 | 4 | 661 | |||
GenBank mRNA | AK005236.1 | - | 4 | 413 | ||
AK011709.1 | - | 4 | 459 | |||
AK082279.1 | - | 4 | 420 | |||
AK082778.1 | - | 4 | 382 | |||
BC013812.1 | - | 4 | 410 | 3483892 | ||
BC054414.1 | - | 4 | 409 | 3257217 | ||
BQ443231.1 | - | 4 | 471 | |||
CD806660.1 | - | 4 | 651 |