Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161825.1 | NP_001155297.1 | 4 | 481 | ||
NM_008822.2 | NP_032848.1 | 5 | 559 | |||
GenBank mRNA | AK004687.1 | - | 5 | 544 | ||
AK160195.1 | - | 5 | 523 | |||
BC132213.1 | - | 5 | 532 | 40130490 | ||
BC132503.1 | - | 5 | 532 | 40130780 | ||
BC145300.1 | - | 4 | 454 | 9053854 | ||
U69171.1 | - | 5 | 550 |