Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162415.1 | NP_001155887.1 | 4 | 354 | ||
NM_001162416.1 | NP_001155888.1 | 2 | 147 | |||
NM_008825.4 | NP_032851.2 | 3 | 248 | |||
GenBank mRNA | AK029659.1 | - | 4 | 355 | ||
AK146744.1 | - | 3 | 242 | |||
BC018418.1 | - | 3 | 205 | 3979400 | ||
BC051014.1 | - | 2 | 161 | 6493944 | ||
EU616666.1 | - | 2 | 146 | |||
EU616667.1 | - | 2 | 146 | |||
EU616668.1 | - | 2 | 146 | |||
EU616669.1 | - | 2 | 146 | |||
EU616670.1 | - | 2 | 156 | |||
X98847.1 | - | 2 | 166 |