Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163487.1 | NP_001156959.1 | 21, 22 | 2233 | ||
NM_001163488.1 | NP_001156960.1 | 21, 22 | 2169 | |||
NM_021514.4 | NP_067489.3 | 20, 21 | 2186 | |||
GenBank mRNA | AF249894.1 | - | 20, 21 | 2126 | ||
AK002711.2 | - | 21, 22 | 2136 | |||
AK049773.1 | - | 21, 22 | 2141 | |||
BC005526.1 | - | 20, 21 | 2150 | 3495173 |