Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021068.2 | NP_066546.1 | 1 | 781 | ||
GenBank mRNA | M27318.1 | - | 1 | 853 |