Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011103.3 | NP_035233.1 | 3 | 367 | ||
GenBank mRNA | AB011812.1 | - | 2 | 130 | ||
AF251036.1 | - | 2 | 130 | |||
AK144231.1 | - | 3 | 372 | |||
AK168768.1 | - | 3 | 362 | |||
AK171476.1 | - | 3 | 368 | |||
AY545076.1 | - | 2 | 130 | |||
BC051416.1 | - | 3 | 331 | 6515302 | ||
HQ840432.1 | - | 2 | 130 | |||
M69042.1 | - | 3 | 362 | |||
X60304.1 | - | 2 | 143 |