Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pklr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099779.1 | NP_001093249.1 | 10 | 1369 | ||
NM_013631.2 | NP_038659.2 | 10 | 1495 | |||
GenBank mRNA | AK149508.1 | - | 9 | 1370 | ||
AK149524.1 | - | 9 | 1368 | |||
BC152327.1 | - | 10 | 1467 | 40141144 | ||
BC152542.1 | - | 10 | 1466 | |||
D63764.1 | - | 10 | 1495 | |||
S79731.1 | - | 10 | 1495 |