Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypla1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008866.2 | NP_032892.1 | 1, 2 | 94 | ||
GenBank mRNA | AK002674.1 | - | 1, 2 | 98 | ||
AK016021.1 | - | 198 | ||||
AK050549.1 | - | 1, 2 | 71 | |||
AK146874.1 | - | 1, 2 | 109 | |||
AK167231.1 | - | 116 | ||||
BC013536.1 | - | 1, 2 | 94 | 4240573 | ||
BC052848.1 | - | 1, 2 | 135 | 30055025 | ||
CT010201.1 | - | 1, 2 | 73 | |||
U89352.1 | - | 1, 2 | 73 |