Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081456.1 | NP_001074925.1 | 15 | 2344 | ||
NM_148937.2 | NP_683739.2 | 16 | 2591 | |||
GenBank mRNA | AK029561.1 | - | 16 | 2604 | ||
AK140231.1 | - | 15 | 2328 | |||
AY033991.1 | - | 16 | 2423 | |||
BC066156.1 | - | 15 | 2235 | 30099811 |